Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53i13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024920.1 | NP_001020091.1 | 4 | 322 | ||
GenBank mRNA | AK213971.1 | - | 3 | 195 | ||
BC147324.1 | - | 4 | 320 | 9056369 | ||
BC147325.1 | - | 4 | 320 | 9056377 | ||
BQ939386.1 | - | 4 | 322 |