Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC030499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287206.1 | NP_001274135.1 | 2 | 298 | ||
GenBank mRNA | BC037104.1 | - | 2 | 236 | ||
BC048862.1 | - | 248 | ||||
BC059191.1 | - | 2 | 247 |