Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172563.3 | NP_766151.1 | 4 | 1395 | ||
GenBank mRNA | AK054062.1 | - | 4 | 885 | ||
AK079068.1 | - | 5 | 856 | |||
AK079308.1 | - | 4 | 770 | |||
BC057693.1 | - | 3 | 816 | 4196259 | ||
BC058705.1 | - | 4 | 1373 | 6839029 |