Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013375.1 | NP_001013393.1 | 9 | 1271 | ||
GenBank mRNA | AK032265.1 | - | 8 | 1798 | ||
AK078033.1 | - | 4 | 407 | |||
AK154637.1 | - | 9 | 1211 | |||
AK167248.1 | - | 9 | 1270 | |||
BC119621.1 | - | 9 | 1244 | 40062238 | ||
BC119622.1 | - | 9 | 1244 | 40062245 |