Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014224.4 | NP_055039.1 | 3 | 320 | ||
GenBank mRNA | BC029055.1 | - | 3 | 299 | 5184316 | |
BC146999.1 | - | 3 | 320 | 9056927 | ||
BC171889.1 | - | 3 | 320 | 9054543 | ||
BC171897.1 | - | 3 | 320 | 9054551 |
Gene Symbol: PGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079807.3 | NP_001073275.1 | 3 | 321 |
Gene Symbol: PGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079808.3 | NP_001073276.1 | 3 | 321 | ||
GenBank mRNA | AK291864.1 | - | 3 | 319 | ||
AK301305.1 | - | 3 | 321 | |||
BC150659.1 | - | 3 | 315 | 9057030 | ||
BC171910.1 | - | 3 | 315 | 9054564 | ||
BC171920.1 | - | 3 | 315 | 9054574 |