Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172261.3 | NP_758465.2 | 1, 2 | 1704 | ||
GenBank mRNA | AY508450.1 | - | 1, 2 | 1704 |