Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146025.2 | NP_666137.1 | 8 | 1609 | ||
GenBank mRNA | AK044765.1 | - | 8 | 1138 | ||
AK047184.1 | - | 8 | 1586 | |||
AK049753.1 | - | 8 | 1612 | |||
BC034054.1 | - | 8 | 1133 | 4505400 | ||
BC049954.1 | - | 8 | 1166 | 4191611 |