Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148953.2 | NP_683755.1 | 2 | 556 | ||
GenBank mRNA | AF403044.1 | - | 2 | 556 | ||
BC127245.1 | - | 2 | 552 | |||
BC127246.1 | - | 2 | 555 | |||
BC152348.1 | - | 2 | 556 | |||
BC152349.1 | - | 2 | 556 | 40104909 |