Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam171a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199200.2 | NP_954670.2 | 7 | 1061 | ||
GenBank mRNA | BC025575.1 | - | 7 | 1037 | ||
BC064455.1 | - | 4 | 794 | 6810415 | ||
CB182176.1 | - | 6 | 809 | |||
CB202938.1 | - | 5 | 527 |