Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199221.2 | NP_954691.2 | 2 | 382 | ||
GenBank mRNA | AB300703.1 | - | 2 | 244 | ||
AK154359.1 | - | 2 | 383 | |||
AK170613.1 | - | 2 | 354 | |||
AY457053.1 | - | 2 | 244 | |||
AY996128.1 | - | 2 | 283 | |||
BC107351.1 | - | 2 | 244 |