Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207012.1 | NP_001193941.1 | 6 | 1030 | ||
NM_001293643.1 | NP_001280572.1 | 7 | 1090 | |||
NM_002632.5 | NP_002623.2 | 7 | 1093 | |||
GenBank mRNA | A18411.1 | - | 6 | 829 | ||
BC001422.2 | - | 7 | 917 | 3139175 | ||
BC007255.1 | - | 7 | 898 | 3028008 | ||
S72960.1 | - | 7 | 583 | |||
X54936.1 | - | 6 | 829 |