Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173755.3 | NP_776116.2 | 15 | 2948 | ||
GenBank mRNA | AK045398.1 | - | 2 | 318 | ||
AK129431.1 | - | 15 | 2948 | |||
BC023329.1 | - | 2 | 678 | |||
BC039233.1 | - | 7 | 1414 | |||
BC051108.1 | - | 6 | 1293 | |||
BC055067.1 | - | 1 | 41 | |||
BC059193.1 | - | 15 | 2573 | 4160667 |