Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167680.1 | NP_001161152.1 | 15 | 1998 | ||
NM_172572.3 | NP_766160.2 | 15 | 2212 | |||
GenBank mRNA | AK028872.1 | - | 15 | 2213 | ||
AK154639.1 | - | 15 | 1979 | |||
AK171085.1 | - | 15 | 1994 | |||
AY167043.1 | - | 15 | 1998 | |||
BC031398.1 | - | 9 | 1031 | |||
BC052182.1 | - | 15 | 1946 | 6335022 |