Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014602.2 | NP_055417.1 | 4 | 1834 | ||
GenBank mRNA | AK314219.1 | - | 4 | 1781 | ||
BC017332.2 | - | 4 | 1799 | |||
BC057239.1 | - | 4 | 1815 | |||
BC110318.1 | - | 4 | 1780 | 5492292 | ||
BC127105.1 | - | 4 | 1651 | 40124707 | ||
BC127106.1 | - | 4 | 1651 | 40124708 | ||
Y08991.1 | - | 4 | 1820 |