Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC017643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252548.1 | NP_001239477.1 | 6 | 659 | ||
NM_001252549.1 | NP_001239478.1 | 6 | 505 | |||
NM_001252550.1 | NP_001239479.1 | 5 | 605 | |||
NM_001254735.1 | NP_001241664.1 | 6 | 582 | |||
NM_144832.2 | NP_659081.1 | 6 | 679 | |||
GenBank mRNA | AK143743.1 | - | 4 | 1696 | ||
AK145884.1 | - | 6 | 645 | |||
AK158402.1 | - | 6 | 616 | |||
AK159453.1 | - | 6 | 637 | |||
AK168104.1 | - | 6 | 637 | |||
AK170767.1 | - | 6 | 627 | |||
BC017643.1 | - | 6 | 635 | 4503381 | ||
BC026616.1 | - | 6 | 570 | 4977003 | ||
BC027406.1 | - | 6 | 464 | 4950930 | ||
BU059230.1 | - | 6 | 608 | |||
CD806773.1 | - | 6 | 582 | |||
CN716786.1 | - | 5 | 531 |