Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153082.4 | NP_694722.2 | 6 | 850 | ||
GenBank mRNA | AK049300.1 | - | 6 | 796 | ||
AK166860.1 | - | 1 | 1335 | |||
AY035893.1 | - | 6 | 637 | |||
BC042643.1 | - | 6 | 765 | 5357595 | ||
BC051140.1 | - | 6 | 812 | 6595101 | ||
CB247441.1 | - | 5 | 564 |