Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144551.5 | NP_653134.2 | 1 | 1449 | ||
GenBank mRNA | AF358867.1 | - | 1 | 314 | ||
AK044747.1 | - | 1 | 1407 | |||
AK080064.1 | - | 1 | 652 | |||
AK082329.1 | - | 1 | 661 | |||
AK144192.1 | - | 1 | 1514 | |||
BC034338.1 | - | 1 | 483 | 5043179 | ||
BC037387.1 | - | 1 | 1393 | 5363174 |