Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161410.1 | NP_001154882.1 | 12 | 2416 | ||
NM_178811.4 | NP_848926.3 | 12 | 2642 | |||
GenBank mRNA | AK049172.1 | - | 11 | 1676 | ||
AK132166.1 | - | 12 | 2428 | |||
AK147985.1 | - | 12 | 2416 | |||
BC002235.1 | - | 4 | 400 | |||
BC024077.1 | - | 12 | 2585 | 5097580 | ||
BC030887.1 | - | 12 | 2410 | 4483611 |