Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199917.1 | NP_001186846.1 | 13 | 1687 | ||
NM_001199918.1 | NP_001186847.1 | 11 | 1345 | |||
NM_001199919.1 | NP_001186848.1 | 12 | 1551 | |||
NM_015599.2 | NP_056414.1 | 12 | 1551 | |||
XM_017010935.1 | XP_016866424.1 | 11 | 1318 | |||
XM_017010936.1 | XP_016866425.1 | 12 | 1550 | |||
XM_017010937.1 | XP_016866426.1 | 10 | 1195 | |||
XM_017010938.1 | XP_016866427.1 | 11 | 1416 | |||
GenBank mRNA | AB032081.1 | - | 12 | 1463 | ||
AF102265.1 | - | 12 | 1467 | |||
AF180371.1 | - | 12 | 1474 | |||
AK001365.1 | - | 11 | 1286 | |||
AK023432.1 | - | 12 | 1496 | |||
AK023709.1 | - | 2 | 177 | |||
AK225363.1 | - | 12 | 1489 | |||
AK301867.1 | - | 13 | 1665 | |||
AK314512.1 | - | 12 | 1484 | |||
AL117443.1 | - | 3 | 262 | |||
BC001258.1 | - | 12 | 1510 | 3451677 |