Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172582.3 | NP_766170.2 | 11 | 1391 | ||
GenBank mRNA | AK077863.1 | - | 11 | 1315 | ||
AK131889.1 | - | 11 | 1353 | |||
AK167380.1 | - | 11 | 1319 | |||
AK170311.1 | - | 10 | 1856 | |||
AK191776.1 | - | 2 | 161 | |||
BC024135.1 | - | 11 | 1292 | |||
BC051055.1 | - | 11 | 1307 | |||
BC057914.1 | - | 8 | 1055 | |||
BC088736.1 | - | 7 | 2640 | |||
BC106183.1 | - | 11 | 1375 | |||
BC132168.1 | - | 11 | 1391 | 40130445 | ||
BC132170.1 | - | 11 | 1391 | 40130447 |