Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172582.3 | NP_766170.2 | 10 | 1282 | ||
GenBank mRNA | AK077863.1 | - | 10 | 1206 | ||
AK131889.1 | - | 10 | 1244 | |||
AK167380.1 | - | 10 | 1210 | |||
AK170311.1 | - | 9 | 1747 | |||
AK191776.1 | - | 1 | 52 | |||
BC024135.1 | - | 10 | 1183 | |||
BC051055.1 | - | 10 | 1198 | |||
BC057914.1 | - | 7 | 946 | |||
BC088736.1 | - | 6 | 2531 | |||
BC106183.1 | - | 10 | 1266 | |||
BC132168.1 | - | 10 | 1282 | 40130445 | ||
BC132170.1 | - | 10 | 1282 | 40130447 |