Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199917.1 | NP_001186846.1 | 13 | 1652 | ||
NM_001199918.1 | NP_001186847.1 | 11 | 1310 | |||
NM_001199919.1 | NP_001186848.1 | 12 | 1516 | |||
NM_015599.2 | NP_056414.1 | 12 | 1516 | |||
XM_017010935.1 | XP_016866424.1 | 11 | 1283 | |||
XM_017010936.1 | XP_016866425.1 | 12 | 1515 | |||
XM_017010937.1 | XP_016866426.1 | 10 | 1160 | |||
XM_017010938.1 | XP_016866427.1 | 11 | 1381 | |||
GenBank mRNA | AF102265.1 | - | 12 | 1432 | ||
AF180371.1 | - | 12 | 1439 | |||
AK001365.1 | - | 11 | 1251 | |||
AK023432.1 | - | 12 | 1461 | |||
AK023709.1 | - | 2 | 142 | |||
AK225363.1 | - | 12 | 1454 | |||
AK301867.1 | - | 13 | 1630 | |||
AK314512.1 | - | 12 | 1449 | |||
AL117443.1 | - | 3 | 227 |