Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161365.1 | NP_001154837.1 | 3 | 376 | ||
NM_177620.4 | NP_808288.2 | 3 | 487 | |||
GenBank mRNA | AK032205.1 | - | 3 | 488 | ||
AK133988.1 | - | 3 | 395 | |||
AK154506.1 | - | 3 | 514 | |||
AK154522.1 | - | 3 | 291 | |||
AK154963.1 | - | 3 | 377 | |||
AK166196.1 | - | 3 | 333 | |||
AK167187.1 | - | 3 | 394 | |||
AK171301.1 | - | 3 | 300 | |||
AK171311.1 | - | 3 | 291 | |||
AK202814.1 | - | 11 | ||||
AY265356.1 | - | 3 | 473 | |||
BC137907.1 | - | 3 | 415 | 8860926 |