Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144834.4 | NP_659083.2 | 3 | 947 | ||
GenBank mRNA | AK050552.1 | - | 924 | |||
AY324633.1 | - | 3 | 947 | |||
BC025821.1 | - | 2 | 909 | 5100400 |