Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198023.2 | NP_932140.1 | 6 | 506 | ||
GenBank mRNA | AB093210.1 | - | 2 | 207 | ||
BC042731.1 | - | 6 | 506 | 4936863 |