Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000926.4 | NP_000917.3 | 4 | 2896 | ||
NM_001202474.3 | NP_001189403.1 | 4 | 2146 | |||
NM_001271162.1 | NP_001258091.1 | 4 | 816 | |||
NR_073141.2 | 4 | 2146 | ||||
NR_073142.2 | 3 | 2029 | ||||
XM_006718858.3 | XP_006718921.1 | 4 | 3746 | |||
GenBank mRNA | AB085843.1 | - | 3 | 2036 | ||
AB085845.1 | - | 4 | 2153 | |||
AF016381.1 | - | 4 | 2328 | |||
AK304853.1 | - | 4 | 816 | |||
AY212933.1 | - | 1 | 1526 | |||
AY382151.1 | - | 3 | 512 | |||
M15716.1 | - | 4 | 2328 | |||
X51730.1 | - | 4 | 3607 |