Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 16 | 1730 | ||
NM_001291444.1 | NP_001278373.1 | 16 | 1736 | |||
NM_001291445.1 | NP_001278374.1 | 15 | 1640 | |||
GenBank mRNA | AK030003.1 | - | 11 | 1099 | ||
AK036581.1 | - | 15 | 1616 | |||
AK054000.1 | - | 16 | 1711 | |||
AK122326.1 | - | 14 | 1419 | |||
BC059076.1 | - | 1 | 300 |