Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160385.2 | NP_001153857.1 | 3 | 950 | ||
NM_001294146.1 | NP_001281075.1 | 3 | 968 | |||
NM_001294154.1 | NP_001281083.1 | 3 | 950 | |||
GenBank mRNA | BB626213.1 | - | 3 | 354 | ||
BU604736.1 | - | 3 | 523 |