Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000927.4 | NP_000918.2 | 16 | 2264 | ||
GenBank mRNA | AB208970.1 | - | 14 | 1734 | ||
AF016535.1 | - | 15 | 1887 | |||
AK290159.1 | - | 16 | 2265 | |||
BC130424.1 | - | 15 | 2096 | 40146455 | ||
EU852583.1 | - | 14 | 1771 | |||
EU854148.1 | - | 13 | 1579 | |||
M14758.1 | - | 16 | 2195 |