Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000443.3 | NP_000434.1 | 3, 4 | 208 | ||
NM_018849.2 | NP_061337.1 | 3, 4 | 208 | |||
NM_018850.2 | NP_061338.1 | 3, 4 | 208 | |||
XM_011516308.2 | XP_011514610.2 | 3, 4 | 232 | |||
XM_011516309.2 | XP_011514611.2 | 3, 4 | 232 | |||
XM_011516310.2 | XP_011514612.2 | 3, 4 | 232 | |||
XM_011516311.2 | XP_011514613.2 | 3, 4 | 232 | |||
XM_011516312.2 | XP_011514614.2 | 3, 4 | 232 | |||
XM_011516313.2 | XP_011514615.1 | 3, 4 | 231 | |||
XM_017012323.1 | XP_016867812.1 | 3, 4 | 219 | |||
GenBank mRNA | BC020618.2 | - | 2, 3 | 139 | ||
BC042531.1 | - | 3, 4 | 244 | 4837724 | ||
M23234.1 | - | 3, 4 | 164 | |||
Z35284.1 | - | 6, 7 | 1981 |