Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146047.2 | NP_666159.2 | 4, 5 | 847 | ||
GenBank mRNA | AK032295.1 | - | 4, 5 | 810 | ||
AK048357.1 | - | 4, 5 | 744 | |||
AK156203.1 | - | 4, 5 | 738 | |||
AK165939.1 | - | 4, 5 | 707 | |||
AK192551.1 | - | 2, 3 | 363 | |||
BC026562.1 | - | 4, 5 | 548 | 5028264 |