Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145452.3 | NP_663427.2 | 21 | 3294 | ||
GenBank mRNA | AK133354.1 | - | 17 | 1854 | ||
AK161497.1 | - | 11 | 1091 | |||
BC013637.1 | - | 19 | 2040 | 3498774 | ||
BC065082.1 | - | 21 | 2836 |