Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170540.1 | NP_001164011.1 | 4 | 484 | ||
NM_145455.3 | NP_663430.2 | 4 | 686 | |||
GenBank mRNA | AK139664.1 | - | 4 | 459 | ||
AK146332.1 | - | 4 | 472 | |||
AK147171.1 | - | 4 | 459 | |||
AK160227.1 | - | 4 | 458 | |||
BC008233.1 | - | 4 | 445 | 2648048 | ||
BC064010.1 | - | 4 | 686 | 30244324 | ||
BC080837.1 | - | 4 | 440 | 5371708 |