Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172592.2 | NP_766180.1 | 3 | 966 | ||
GenBank mRNA | AK028794.1 | - | 1 | 518 | ||
AK033354.1 | - | 3 | 966 | |||
AK037115.1 | - | 3 | 942 | |||
AK039311.1 | - | 5 | 701 | |||
AK044086.1 | - | 2 | 270 | |||
AK052265.1 | - | 5 | 835 | |||
AK054414.1 | - | 3 | 1417 | |||
BC063761.1 | - | 6 | 711 | |||
BC070460.1 | - | 3 | 339 | 30548562 |