Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178683.4 | NP_848798.1 | 5, 6 | 789 | ||
GenBank mRNA | AK036550.1 | - | 5, 6 | 748 | ||
AK043283.1 | - | 746 | ||||
AK077676.1 | - | 5, 6 | 790 | |||
AK083480.1 | - | 5, 6 | 785 | |||
BC152330.1 | - | 5, 6 | 794 | 40141538 | ||
BC152331.1 | - | 5, 6 | 794 | 40141540 |