Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139299.2 | NP_647460.2 | 5, 6 | 591 | ||
GenBank mRNA | AB083111.1 | - | 5, 6 | 590 | ||
AF486621.1 | - | 2, 3 | 170 | |||
AY509150.1 | - | 487 | ||||
AY509151.1 | - | 487 | ||||
BC066164.1 | - | 3, 4 | 312 | 30475340 |