Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139299.2 | NP_647460.2 | 11, 12 | 1532 | ||
GenBank mRNA | AB083111.1 | - | 11, 12 | 1531 | ||
AF486621.1 | - | 8, 9 | 1111 | |||
AY509150.1 | - | 1428 | ||||
AY509151.1 | - | 1428 | ||||
BC066164.1 | - | 9, 10 | 1253 | 30475340 | ||
BC145083.1 | - | 10, 11 | 1435 | |||
BC145084.1 | - | 10, 11 | 1435 | |||
BC145085.1 | - | 11, 12 | 1558 |