Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081062.1 | NP_001074531.1 | 1 | 421 | ||
GenBank mRNA | AK053890.1 | - | 1 | 420 | ||
BC147759.1 | - | 1 | 397 | 9087484 | ||
BC147766.1 | - | 1 | 397 | 9087491 |