Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3830406C13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284383.1 | NP_001271312.1 | 4 | 290 | ||
NM_001284384.1 | NP_001271313.1 | 4 | 366 | |||
NM_001284385.1 | NP_001271314.1 | 4 | 369 | |||
NM_001284386.1 | NP_001271315.1 | 3 | 209 | |||
NM_001284387.1 | NP_001271316.1 | 3 | 209 | |||
NM_146051.3 | NP_666163.1 | 4 | 293 | |||
NM_178141.3 | NP_835277.1 | 4 | 293 | |||
GenBank mRNA | AK076182.1 | - | 4 | 284 | ||
AK089971.1 | - | 4 | 266 | |||
BC027421.1 | - | 3 | 253 | 4194414 | ||
BC057656.1 | - | 4 | 315 | 4020532 | ||
BC094456.1 | - | 4 | 271 | 5137611 | ||
BE624571.1 | - | 2 | 169 | |||
BQ715343.1 | - | 3 | 251 | |||
BY233185.1 | - | 4 | 279 |