Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033270.2 | NP_001028442.2 | 12 | 1688 | ||
GenBank mRNA | AK049322.1 | - | 3 | 350 | ||
AK088400.1 | - | 12 | 1610 | |||
AK152233.1 | - | 12 | 1689 | |||
AK153567.1 | - | 12 | 1689 | |||
BC038373.1 | - | 13 | 1581 | |||
GU386352.1 | - | 12 | 1595 | |||
GU386353.2 | - | 12 | 1634 | |||
HM624050.1 | - | 12 | 1498 | |||
HM624051.1 | - | 13 | 1828 | |||
JQ073566.1 | - | 12 | 1480 | |||
JQ344322.1 | - | 12 | 1459 | |||
JQ347261.1 | - | 13 | 1558 | |||
JQ349031.1 | - | 13 | 1694 | |||
JQ349032.1 | - | 13 | 1655 | |||
JQ349033.1 | - | 13 | 1519 | |||
JX254908.1 | - | 13 | 2003 | |||
JX254909.1 | - | 13 | 1964 | |||
JX254910.1 | - | 13 | 2003 | |||
JX254911.1 | - | 13 | 1964 | |||
JX254912.1 | - | 13 | 1867 | |||
JX310364.1 | - | 13 | 1964 | |||
KF279520.1 | - | 14 | 1598 | |||
KF279521.1 | - | 14 | 2034 |