Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033270.2 | NP_001028442.2 | 10 | 1446 | ||
GenBank mRNA | AK049322.1 | - | 1 | 108 | ||
AK088400.1 | - | 10 | 1368 | |||
AK152233.1 | - | 10 | 1447 | |||
AK153567.1 | - | 10 | 1447 | |||
BC038373.1 | - | 11 | 1339 | |||
GU386352.1 | - | 10 | 1353 | |||
GU386353.2 | - | 10 | 1392 | |||
HM624050.1 | - | 10 | 1256 | |||
HM624051.1 | - | 11 | 1586 | |||
JQ073566.1 | - | 10 | 1238 | |||
JQ344322.1 | - | 10 | 1217 | |||
JQ347261.1 | - | 11 | 1316 | |||
JQ349031.1 | - | 11 | 1452 | |||
JQ349032.1 | - | 11 | 1413 | |||
JQ349033.1 | - | 11 | 1277 | |||
JX254908.1 | - | 11 | 1761 | |||
JX254909.1 | - | 11 | 1722 | |||
JX254910.1 | - | 11 | 1761 | |||
JX254911.1 | - | 11 | 1722 | |||
JX254912.1 | - | 11 | 1625 | |||
JX310364.1 | - | 11 | 1722 | |||
KF279520.1 | - | 12 | 1356 | |||
KF279521.1 | - | 12 | 1792 |