Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136240.1 | NP_001129712.1 | 7 | 1648 | ||
NM_172264.2 | NP_758468.2 | 7 | 1446 | |||
NM_175343.5 | NP_780552.1 | 7 | 1621 | |||
GenBank mRNA | AK030900.1 | - | 7 | 1622 | ||
AK052715.1 | - | 7 | 1629 | |||
AK087570.1 | - | 7 | 1557 | |||
AK164042.1 | - | 7 | 1484 | |||
BC039186.1 | - | 7 | 1446 | 3979709 |