Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005392.3 | NP_005383.3 | 13, 14 | 1964 | ||
XM_005252051.2 | XP_005252108.1 | 13, 14 | 1958 | |||
XM_006717143.2 | XP_006717206.1 | 13, 14 | 1853 | |||
GenBank mRNA | AB014562.2 | - | 12, 13 | 1857 | ||
AK294861.1 | - | 1, 2 | 102 | |||
AL834263.1 | - | 13, 14 | 1815 | |||
BC028051.2 | - | 1, 2 | 72 | |||
BC152437.1 | - | 12, 13 | 1857 |