Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144553.2 | NP_653136.2 | 6 | 960 | ||
GenBank mRNA | AB076696.1 | - | 6 | 960 | ||
AK036384.1 | - | 6 | 762 | |||
AK048470.1 | - | 6 | 730 | |||
AK085882.1 | - | 6 | 741 | |||
AK129032.1 | - | 6 | 706 | |||
AK135820.1 | - | 6 | 767 | |||
AK147955.1 | - | 6 | 730 | |||
BC027273.1 | - | 6 | 700 | |||
BC058087.1 | - | 6 | 694 | 6816660 |