Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144553.2 | NP_653136.2 | 2 | 476 | ||
GenBank mRNA | AB076696.1 | - | 2 | 476 | ||
AK036384.1 | - | 2 | 278 | |||
AK048470.1 | - | 2 | 246 | |||
AK085882.1 | - | 2 | 257 | |||
AK129032.1 | - | 2 | 222 | |||
AK135820.1 | - | 2 | 283 | |||
AK147955.1 | - | 2 | 246 | |||
AK164575.1 | - | 2 | 271 | |||
BC027273.1 | - | 2 | 216 | |||
BC058087.1 | - | 2 | 210 | 6816660 |