Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172599.4 | NP_766187.1 | 6, 7 | 908 | ||
GenBank mRNA | AK029330.1 | - | 6, 7 | 889 | ||
AK029967.1 | - | 6, 7 | 889 | |||
BC057360.1 | - | 5, 6 | 628 | |||
BC090995.1 | - | 6, 7 | 888 | 6754513 |