Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172600.3 | NP_766188.3 | 11 | 1337 | ||
GenBank mRNA | AK031240.1 | - | 12 | 1523 | ||
AK032809.1 | - | 11 | 1308 | |||
AK038175.1 | - | 11 | 1293 | |||
AK040456.1 | - | 10 | 1118 | |||
AK054494.1 | - | 11 | 1310 | |||
AK141246.1 | - | 11 | 1343 | |||
AK211943.1 | - | 2 | 161 | |||
BC046454.1 | - | 3 | 207 | |||
BC046491.1 | - | 3 | 207 | |||
BC059217.1 | - | 11 | 1113 | 6408599 |