Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080949.2 | NP_001074418.1 | 4 | 490 | ||
NM_177625.4 | NP_808293.1 | 4 | 490 | |||
GenBank mRNA | AK044781.1 | - | 4 | 480 | ||
AK077874.1 | - | 3 | 811 | |||
AK081214.1 | - | 4 | 466 | |||
AK133953.1 | - | 4 | 472 | |||
AK157777.1 | - | 4 | 469 | |||
AK161279.1 | - | 4 | 468 | |||
BC003272.1 | - | 4 | 460 | |||
BC025610.1 | - | 4 | 472 | 5257921 | ||
BC069946.1 | - | 4 | 471 | |||
BC092074.1 | - | 4 | 464 | 4920436 |