Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080949.2 | NP_001074418.1 | 2 | 105 | ||
NM_177625.4 | NP_808293.1 | 2 | 105 | |||
GenBank mRNA | AK044781.1 | - | 2 | 95 | ||
AK077874.1 | - | 2 | 82 | |||
AK081214.1 | - | 2 | 81 | |||
AK157777.1 | - | 2 | 84 | |||
AK161279.1 | - | 2 | 83 | |||
AK207059.1 | - | 2 | 41 | |||
BC003272.1 | - | 2 | 75 | |||
BC025610.1 | - | 2 | 87 | 5257921 | ||
BC069946.1 | - | 2 | 86 | |||
BC092074.1 | - | 2 | 79 | 4920436 | ||
BY278375.1 | - | 2 | 105 |