Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172603.3 | NP_766191.1 | 2 | 178 | ||
GenBank mRNA | AK077176.1 | - | 2 | 173 | ||
BC116758.1 | - | 2 | 162 | 40126077 | ||
BC116760.1 | - | 2 | 162 | 40126079 | ||
BY162942.1 | - | 2 | 178 | |||
EU155109.1 | - | 2 | 162 |